[EN]KLERCIDE SP/ICIDAL LOW RES PEROXIDE 1 * 4 KS
Catalog # SHIE6600543
Supplier: ECOLAB CONTAMINATION CONTROL
[EN]KLERCIDE SP/ICIDAL LOW RES PEROXIDE 1 * 4 KS
Catalog # SHIE6600543
Supplier: ECOLAB CONTAMINATION CONTROL
Frequently Bought Together
Isooktán ≥99.8% (by GC), SPECTRONORM® for spectroscopy
Supplier: VWR Chemicals
Ethylacetát ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Toluen, SupraSolv® for GC-ECD/FID, Supelco®
Supplier: Merck
Voda USP, JP, Ph. Eur., purified
Supplier: VWR Chemicals
Cyklohexan, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Dimethylsulfoxid ≥99.5% (by GC), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethylacetát ACS, ISO, Reag. Ph. Eur., Burdick & Jackson™
Supplier: Honeywell Chemicals
[EN]KLERCIDE SP/CIDAL LOW RES PEROXIDE ( 1 * 6 KS
Supplier: ECOLAB CONTAMINATION CONTROL
Xylen (směs izomerů), BAKER, J.T.Baker®
Supplier: Avantor
Toluen 99.85% for HPLC
Supplier: Thermo Fisher Scientific
Acetonitril, extra dry 99.9%, AcroSeal®
Supplier: Thermo Fisher Scientific
Dimethylsulfoxid 99.8+% for HPLC
Supplier: Thermo Fisher Scientific
Xylen (směs izomerů) ≥99% (isomers plus ethylbenzene)
Supplier: Honeywell Chemicals
Izobutanol 99+% for HPLC
Supplier: Thermo Fisher Scientific