[EN]KLERCIDE SPORICIDAL LOW RESIDUE PERO 1 * 6 KS
Katalogové číslo SHIE6600539
Dodavatel: ECOLAB CONTAMINATION CONTROL
[EN]KLERCIDE SPORICIDAL LOW RESIDUE PERO 1 * 6 KS
Katalogové číslo SHIE6600539
Dodavatel: ECOLAB CONTAMINATION CONTROL
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
Dodavatel: Dr. Weigert
Plynové kartuše, Campingaz®
Dodavatel: CAMPING GAZ
VWR®, pH a redoxní elektrody univerzální
Dodavatel: VWR Collection
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
Chlorid sodný, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Baka kuelov dle Erlenmeyera s NZ 29/32, 100ml 1 * 1 KS
Dodavatel: GLASSCO