[EN]KLERCIDE SP/CIDAL LOW RES PEROXIDE ( 1 * 6 KS
Catalog # SHIE6600535
Supplier: ECOLAB CONTAMINATION CONTROL
[EN]KLERCIDE SP/CIDAL LOW RES PEROXIDE ( 1 * 6 KS
Catalog # SHIE6600535
Supplier: ECOLAB CONTAMINATION CONTROL
Frequently Bought Together
Bezpečnostní brýle, uvex astrospec 2.0
Supplier: UVEX
Aceton, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Supplier: Merck
Cyklohexan ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
Ethylacetát ≥99%, TECHNICAL
Supplier: VWR Chemicals
Methanol, LiChrosolv® for liquid chromatography, Supelco®
Supplier: Merck
Voda, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
KLERCIDE LOW RESIDUE QUAT 1L WFI STERILE 1 * 6 KS
Supplier: ECOLAB CONTAMINATION CONTROL
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Petrolether, 40…60 °C, GPR RECTAPUR®
Supplier: VWR Chemicals
[EN]KLERCIDE SPORICIDAL LOW RESIDUE PERO 1 * 4 KS
Supplier: ECOLAB CONTAMINATION CONTROL
Alcohols, Sterile, Klercide™ 70/30 IPA Blended with WFI and Klercide™ 70/30 Denatured Ethanol Blended with WFI
Supplier: ECOLAB CONTAMINATION CONTROL