[EN]CHLOROTHALONIL METABOLITE R417888 SO 1 * 5 mg
Catalog # EHERC11510455
Supplier: EHRENSTORFER
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Supplier: VWR Chemicals
pH buffers, ready to use
Supplier: WTW
Capric Alcohol, Reference Standard
Supplier: USP
VWR®, Flow Cup viscosity standards
Supplier: VWR Chemicals
CIPROFLOXACIN FORMAMIDE 1 * 200 mg
Supplier: Merck
[EN]NISTmAb, HUMANIZED IGG1K MONICLONAL 1 * 1 KS
Supplier: Merck
Buffer Solutions
Supplier: Thermo Fisher Scientific
SULFITE LR TABLETS 1 * 100 Tablet
Supplier: Lovibond
WEDELOLACTONE 1 * 15 mg
Supplier: EDQM
TESTOSTERONE PROPIONATE CERTIFIED 1 * 0,1 g
Supplier: EHRENSTORFER
pH Standard Solutions, IUPAC, Certified
Supplier: Hach
PCB-MIX 1 10 NG/µL 1 * 10 mL
Supplier: EHRENSTORFER