4'-NITRO-[1,1'-BIPHENYL]-4-CARBOXYLIC AC 1 * 5 g
Katalogové číslo APOSOR315357-5G
Dodavatel: Apollo Scientific
4'-NITRO-[1,1'-BIPHENYL]-4-CARBOXYLIC AC 1 * 5 g
Katalogové číslo APOSOR315357-5G
Dodavatel: Apollo Scientific
Frequently Bought Together
Dimethylsulfoxid, dehydrated (max. 0.03% H₂O) ≥99.9%, AnalaR NORMAPUR® ACS analytical reagent
Dodavatel: VWR Chemicals
Petrolether, 60…80 °C, EMSURE® p.a., Supelco®
Dodavatel: Merck
Dimethylsulfoxid, extra dry over Molecular Sieve 99.7+%, AcroSeal®
Dodavatel: Thermo Fisher Scientific
Glycerin ≥99%
Dodavatel: Thermo Fisher Scientific
Toluen ≥99.8% (by GC), ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Dodavatel: MACRON AVANTOR BRAND
Methanol 99.8% for HPLC
Dodavatel: Thermo Fisher Scientific
Chloroform ≥99.5% stabilizovaný
Dodavatel: Honeywell Chemicals
[EN]ETHYL ACETATE EXTRA PURE, DAC, N. F. 1 * 100 L
Dodavatel: Honeywell Chemicals
Tetramethylamoniumhydroxid 25% (w/w) in methanol
Dodavatel: Thermo Fisher Scientific
Pyridin, extra dry over Molecular Sieve 99.5%, AcroSeal®
Dodavatel: Thermo Fisher Scientific
n-Pentan, anhydrous 99+%, extra pure
Dodavatel: Thermo Fisher Scientific
Dodecyltrimethylammonium bromide, Supelco®
Dodavatel: Merck
ethylbenzen 99.8%, pure
Dodavatel: Thermo Fisher Scientific