[EN]QUARTER-WAVE PLATE F. COMPENSATOR SL 1 * 1 KS
Catalog # LEIM11553387
Supplier: LEICA MICROSYSTEMS
[EN]QUARTER-WAVE PLATE F. COMPENSATOR SL 1 * 1 KS
Catalog # LEIM11553387
Supplier: LEICA MICROSYSTEMS
Frequently Bought Together
Gas detection tubes for water vapour
Supplier: Dräger
Single gas detectors, BW Clip
Supplier: BW TECHNOLOGIES
Accessories for OxiTop® and OxiTop® Control
Supplier: WTW
VWR® Counter-Pen™ Cell Counter
Supplier: VWR Collection
Components for spectrophotometric colorimeters, PFX<i>i</i>-195/880/950/995 series, Lovibond®
Supplier: Lovibond Tintometer
VWR® P9, Spektrofotometr UV/Vis
Supplier: VWR Collection
Accessories for VWR® V-1200, UV-1600PC, UV-3100PC
Supplier: VWR Collection
Accessories for Mettler balances
Supplier: Mettler - Toledo
Bellows pumps, accuro®
Supplier: Dräger
Conductivity standards
Supplier: WTW
VWR®, Square Cuvettes
Supplier: VWR Collection
VWR® pHenomenal®, Stojan na elektrody
Supplier: VWR Collection
VWR®, Lupy zvětšovací s osvětlením
Supplier: VWR Collection
Titration beakers
Supplier: Mettler - Toledo
pH/mV/°C meters, handheld, pH 3110 / 3310
Supplier: WTW
Flongle Adapter and Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Flow Cells for MinION™ and GridION™ Devices
Supplier: Oxford Nanopore Technologies
Accessories for Büchi melting point apparatus B540 and B545
Supplier: BUCHI
Accessories for CytoFLEX Platform
Supplier: Beckman Coulter