[EN]SODIUM HYDROXIDE SOLUTION 50% IN H2O 1 * 500 mL
Katalogové číslo SIAL415413-500ML
Dodavatel: Merck
[EN]SODIUM HYDROXIDE SOLUTION 50% IN H2O 1 * 500 mL
Katalogové číslo SIAL415413-500ML
Dodavatel: Merck
Frequently Bought Together
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Dodavatel: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Dodavatel: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Dodavatel: Merck
Test indikátor pH proužky, pH-Fix
Dodavatel: MACHEREY-NAGEL
Hydroxid sodný, pellets, EMSURE® p.a., Supelco®
Dodavatel: Merck
Immersion oil for microscopy, Sigma-Aldrich®
Dodavatel: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Dodavatel: Merck
Kyselina trifluoroctová ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Mravenčan amonný ≥99%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Test pro stanovení peroxidu, MQuant™, Supelco®
Dodavatel: Merck
Test kits, photometric, COD, LCK cuvette tests
Dodavatel: Hach