[EN]FE 1000MG/L IN DILUTED HNO3 FOR ICP/ 1 * 100 mL
Catalog # ACSDM219.2NP.L1
Supplier: CPACHEM
Frequently Bought Together
Test kits, phosphate, Spectroquant®, Supelco®
Supplier: Merck
Hydroxid sodný, pellets, EMSURE® p.a., Supelco®
Supplier: Merck
Kyselina trifluoroctová ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Qualitative test paper phosphatesmo MI for phosphatase in milk
Supplier: MACHEREY-NAGEL
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Chlornan sodný (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
Formaldehyde 4% stabilised, buffered (pH 7,0 ±0,1)
Supplier: VWR Chemicals
DPD NO.1 REAGENT SOLUTION 1 * 100 mL
Supplier: Lovibond
Formalin 10%, Q Path®, buffered
Supplier: VWR Chemicals
Test kits, manganese, Spectroquant®, Supelco®
Supplier: Merck
Schiff reagent for histology, Sigma-Aldrich®
Supplier: Merck
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Hydroxid sodný 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Supplier: VWR Chemicals
Lugol's iodine solution, Sigma-Aldrich®
Supplier: Merck
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach