Orders LinkContactUsLinkComponent
VeriFluor™ Far-Red Passive Reference Dye
02, VeriFluor™ far-red passive reference dye, 400× in water, 640/660 nm, 500 µl
  29087-500UL
 :   Biotium
VeriFluor™ Far-Red Passive Reference Dye
  29087-500UL
 :  Biotium
 :  
VeriFluor™ Far-Red Passive Reference Dye
02, VeriFluor™ far-red passive reference dye, 400× in water, 640/660 nm, 500 µl
 :  
30 330,00 Kč
1 * 500 µl
A far-red passive reference dye for qPCR that frees up the ROX channel for use with JUN™ probes, EvaRuby™ qPCR dye, and other orange dyes.
  • An alternative to ROX for qPCR normalization in the red channel
  • Drop-in replacement for Mustang Purple™ on most instruments
  • Tolerates DTT in qPCR reactions, unlike Mustang Purple™
  • More soluble than ROX for easier mixing and more accurate results
  • Stand-alone dye solution for use with our Forget-Me-Not™ Master Mixes or other mixes
VeriFluor™ Far-Red is a far-red passive reference dye that is used to normalize the fluorescent reporter signal during qPCR and eliminate well-to-well variation. It is an alternative to normalization with ROX passive reference dye and a direct replacement for Mustang Purple™.
  • Bal.:
    500 µl
  • Popis:
    VeriFluor™ far-red passive reference dye, 400× in water
  • Sekvence:
    02
  • Ex/Em:
    640/660 nm
  • Cat. No.:
    29087-500UL
  • Podmínky uskladnění:
    Store at –10...–35 °C