Orders LinkContactUsLinkComponent
Erlenmeyer Flasks with Screw Neck
Erlenmeyer Flasks with Screw Neck
2, 50 ml, Jantarová, GL 25
Katalogové číslo LENZ81004050
Dodavatel:   Lenz Laborglas
Erlenmeyer Flasks with Screw Neck
Katalogové číslo LENZ81004050
Dodavatel:  Lenz Laborglas
Erlenmeyer Flasks with Screw Neck
2, 50 ml, Jantarová, GL 25
Přejít na:
Price
1 960,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
DURAN®, amber coloured, with DIN thread.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to rapid temperature changes
With white volume graduations and white marking field. Amber coloured to protect light-sensitive substances.
Specifikace
  • Závit:
    GL 25
  • Kapacita:
    50 ml
  • Barva:
    Jantarová
  • Sekvence:
    2