Pipette tips
51, Standard, 1000 - 5000 µl, -, Transparent, Kartell®, Eppendorf®, Biohit®, Socorex® , HTL®, Rack
Katalogové číslo KART0964900
Dodavatel: KARTELL
Přejít na:
Product Details & Documents
These tips are recommended for use with the Kartell automatic micropipettes but deliver excellent results even if used with the most other micropipette brands available on the market.
- High clarity and excellent water-repellent properties designed to keep sample retention within the tip to a minimum
- Precise centering of the orifice at the end of the tip guarantees reliable directionality of the sample when delivered
- Tips in racks come with snap-on lid
- Boxes containing 'macro' tips (5000 μl and 10000 μl) have a sliding horizontal cover
- Containers and tips are autoclavable to +121 °C for 20 minutes
Specifikace
- Balené:Rack
- Sterilní:-
- Objem:1000 - 5000 µl
- Sekvence:51
- Druh špičky:Standard
- Barva špičky:Transparent
- Pro použití s:Kartell®, Eppendorf®, Biohit®, Socorex® , HTL®
- Cat. No.:KART0964900