Orders LinkContactUsLinkComponent
Micro Test Tubes, Graduated
Micro Test Tubes, Graduated
04, Micro test tube, graduated, 1,5 ml, Růžová, PP
Katalogové číslo KART0027913
Dodavatel:   KARTELL
Micro Test Tubes, Graduated
Katalogové číslo KART0027913
Dodavatel:  KARTELL
Micro Test Tubes, Graduated
Micro Test Tubes, Graduated
04, Micro test tube, graduated, 1,5 ml, Růžová, PP
Přejít na:
Price
5 750,00 Kč
Přihlásit se pro dostupnost
1 * 10.000 KS
Product Details & Documents
Ideal for samples analysis.
  • Made from PP
  • Flat cap with writing area
  • Available in blue, yellow, pink and green
Depending on centrifuge being used, these tubes can withstand a centrifugal force of up to RCF 11000.
Certifications: The raw materials with the relevant requirements of Food-contact EU; Regulation 1935/2004 EC, Regulation 2023/2006/EC, FDA and REACH/ROHS regulations.
Specifikace
  • Kapacita:
    1,5 ml
  • Barva:
    Růžová
  • Popis:
    Micro test tube, graduated
  • Materiál:
    PP
  • Sekvence:
    04