n-Hexan ≥99.0%, SPECTRONORM® for spectroscopy
2725, Glass bottle, 500 mL
Dodavatel: VWR Chemicals
n-Hexan ≥99.0%, SPECTRONORM® for spectroscopy
Katalogové číslo 140095D
Dodavatel: VWR Chemicals
Číslo CAS: 110-54-3
Přejít na:
Product Details & Documents
SPECTRONORM solvents have extensive specifications to meet the highly demanding requirements of UV applications.
- Meets the requirements of "Specific use spectroscopy" according Reag. Ph.Eur.
Synonyma: Hexyl hydride, Hexanen, Hexane
- Číslo CAS:110-54-3
- Vzorec:H₃C(CH₂)₄CH₃
- MDL číslo:MFCD00009520
- Molecular Weight:86.18
- Merck Index:13,04712
- Bod varu:69 °C (1013 hPa)
- Bod tání:–94,3 °C
- Hustota:0,659 g/cm³ (20 °C)
- Bod vzplanutí:–22 °C
- Teplota pro skladování:Ambient
- EINECS:203-777-6
- UN:1208
- ADR:3,II
- :Filtered through a 0.2 µm filter, packaged under nitrogen.
- :Density: 0,659 g/cm³ (20 °C)
- REACH:01-2119480412-44
Documents
Certifikáty
Hledat podle čísla šarže2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
P20_VWRC140095D_SPEC_EN.pdf spec_VWRC140095D spec_VWRC140096E
Product specification
Specifikace
Výsledky testů specifikací
- Assay (GC):≥ 99.0 %
- Acidity:≤ 0.0002 meq/g
- Alkalinity:≤ 0.0002 meq/g
- Colouration:≤ 10 APHA
- Evaporation residue:≤ 2 ppm
- Water:≤ 50 ppm
- Absorbance (195 nm):≤ 1.00
- Absorbance (210 nm):≤ 0.22
- Absorbance (217 nm):≤ 0.10
- Absorbance (225 nm):≤ 0.05
- Absorbance (from 245 nm):≤ 0.01
- Transmittance (195 nm):≥ 10 %
- Transmittance (210 nm):≥ 60 %
- Transmittance (217 nm):≥ 80 %
- Transmittance (225 nm):≥ 90 %
- Transmittance (from 245 nm):≥ 98 %
- Fluorescence (as quinine) (254 nm):≤ 1.0 ppb
- Fluorescence (as quinine) (365 nm):≤ 1.0 ppb
Specifikace
- Typ balení:Glass bottle
- Bal.:500 mL
- Sekvence:2725
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



