[EN]CRYOBOX PP 81 PLACES 3-5ML NATURAL 1 * 5 KS
Catalog # RATI5120020
Supplier: RATIOLAB
Frequently Bought Together
Nálevka s krátkým stonkem,úhel 60°,75mm 1 * 1 KS
Supplier: Kavalier
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Supplier: EPPENDORF
VWR®, Microcentrifuge Tubes, High Performance
Supplier: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection
VWR®, Misky Petriho, PS
Supplier: VWR Collection
VWR®, Zkumavky pro PCR, 0,2 ml
Supplier: VWR Collection
Mikrocentrifugační zkumavky Eppendorf® Protein LoBind®
Supplier: EPPENDORF
VWR®, Sérologická pipeta, standardní
Supplier: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Supplier: VWR Collection
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Supplier: VWR Collection
VWR®, Pipety Pasteurovy, Standard Line
Supplier: VWR Collection