Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]TASKI SPRINT MULTI 6X0.5L 1 * 3 L
Catalog # DIVY101106371
Supplier:
Diversey
Expand image
[EN]TASKI SPRINT MULTI 6X0.5L 1 * 3 L
Catalog # DIVY101106371
Supplier:
Diversey
Frequently Bought Together
VWR®, Nitrile Extra Light, Rukavice nitrilové
Supplier: VWR Collection
VWR®, Popisovače laboratorní
Supplier: VWR Collection
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Ubrousky čisticí
Supplier: VWR Collection
Rukavice, nitril/neopren, Microflex® 93-260
Supplier: Ansell
Rukavice, nitrilové, TouchNTuff® 92-600 a 92-605
Supplier: Ansell
Rukavice, nitrilové, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR®, Latexové rukavice
Supplier: VWR Collection
VWR® NITRILE LIGHT, Rukavice, nitrilové
Supplier: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Supplier: VWR Collection