Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
n-Hexan for gas chromatography, Burdick & Jackson™

n-Hexan for gas chromatography, Burdick & Jackson™

5096, Glass bottle, 2,5 L
Catalog # HONC65747-2.5L
Flammable
Health
Irritant
Environment
Supplier:   Honeywell Chemicals
n-Hexan for gas chromatography, Burdick & Jackson™
Catalog # HONC65747-2.5L
CAS Number:  110-54-3
n-Hexan for gas chromatography, Burdick & Jackson™
n-Hexan for gas chromatography, Burdick & Jackson™

5096, Glass bottle, 2,5 L

CAS Number:  110-54-3
Price
4 680,00 Kč
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Hexyl hydride, Hexanen, Hexane
  • CAS Number:110-54-3
  • Formula:H₃C(CH₂)₄CH₃
  • MDL Number:MFCD00009520
  • Molecular Weight:86.18
  • Boiling Pt:66…69 °C (1,013 hPa)
  • Melting Pt:–95 °C
  • Density:0,660…0.668 g/cm3 (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-777-6
  • UN:1208
  • ADR:3,II
  •  :Density: 0,659 g/cm³ (20 °C)
Specifications

Specification Test Results

  • Assay (GC):
    Min. 95.0 %
  • Non-volatile matter:
    Max. 0.0003 %
  • Water (Karl Fischer):
    Max. 0.01 %
  • Suitability for residue analysis:
    Max. 5 ng/l

Specifications

  • Typ balení:
    Glass bottle
  • Bal.:
    2,5 L
  • Sekvence:
    5096

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.