[EN]2-METHYL-1-PYRROLINE CAS 872-32-2 UN 1 * 25 g
Katalogové číslo ABCRAB204846
Dodavatel: abcr
Frequently Bought Together
1-Propanol ≥99%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Ethylacetát ≥99.5% for HPLC
Dodavatel: Thermo Fisher Scientific
Chloroform ≥99.5% stabilizovaný
Dodavatel: Honeywell Chemicals
Dichlormethan 99.8% stabilised for HPLC
Dodavatel: Thermo Fisher Scientific
n-Hexan 95% for residue analysis, for trace analysis of polyaromatic hydrocarbons
Dodavatel: Thermo Fisher Scientific
Aceton 99.8% for HPLC
Dodavatel: Thermo Fisher Scientific
Cyklohexan for HPLC
Dodavatel: Thermo Fisher Scientific
Cyklohexan ≥99%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Diisopropylether stabilizovaný p.a.
Dodavatel: Thermo Fisher Scientific
Aceton ≥99.5% ACS, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
N,N-Dimethylacetamid, anhydrous (max. 0.005% H₂O)
Dodavatel: VWR Chemicals