Masterflex® Fitting, PVDF, Tee, Hose Barb Reducer, 1/4" ID x 1/4" ID x 1/8" ID; 10/PK
Katalogové číslo MFLX40614-36
Dodavatel: Avantor Fluid Handling
O tomto produktu
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Velikost ozubu/vnit. Ø hadičky (port 1):1/4"
- Velikost ozubu/vnit. Ø hadičky (port 2):1/4"
- Typ připojení:Barbed Reducer
- Flow pattern:T
- Popis:Fitting, PVDF, Tee, Hose Barb Reducer, 1/4" ID x 1/4" ID x 1/8" ID; 10/PK
- Environmentally Preferable:
- Sterilizace:Autoclave or ethylene oxide (ETO)
- Bal.:10
- Barb size/tubing int.Ø (port 3):1/8"
- Fitting material:PVDF
Frequently Bought Together