CHO K1 MOUSE CD38 STABLE CELL LINE 1 * 2 Vial
Katalogové číslo GENSM00636
Dodavatel: Genscript
Frequently Bought Together
Eppendorf Tubes® Microtubes with Snap Cap, 5,0 ml
Dodavatel: EPPENDORF
Mikrocentrifugační zkumavky Eppendorf® Protein LoBind®
Dodavatel: EPPENDORF
VWR® Water Sampling Bottles, with Tamper Evident Cap
Dodavatel: VWR Collection
VWR®, Zkumavky pro PCR, 0,2 ml
Dodavatel: VWR Collection
Cell culture flasks, Falcon®
Dodavatel: Corning
VWR®, Pipety Pasteurovy, Standard Line
Dodavatel: VWR Collection
Test tubes
Dodavatel: Greiner Bio-One
VWR®, Centrifuge Tubes, Conical Bottom, with Flat Screw Cap
Dodavatel: VWR Collection
Eppendorf® DNA LoBind® Microcentrifuge Tubes
Dodavatel: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
Dodavatel: EPPENDORF
VWR®, Sérologická pipeta, standardní
Dodavatel: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Dodavatel: Corning
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Dodavatel: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Dodavatel: EPPENDORF
Accessories and consumables for KingFisher™ Flex
Dodavatel: Thermo Fisher Scientific