PROTEIN G RESIN FF PREPACK COLUMN 5ML X5 1 * 5 mL
Catalog # GENSL00681-55
Supplier: Genscript
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Supplier: EPPENDORF
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Supplier: VWR Collection
Petri dish racks, stainless steel, PetriPile®
Supplier: INTERSCIENCE
VWR®, Ice Pans, with or without Lid
Supplier: VWR Collection
Serological pipettes, Stripette™
Supplier: Corning
Containers, straight, wide mouth, with screw cap, Corning® Gosselin™
Supplier: Corning
VWR®, Misky kultivační, vícejamkové
Supplier: VWR Collection
Deepwell Plates 96-Well, 2000 µl
Supplier: EPPENDORF
Volumetric measuring scoops, SteriWare®
Supplier: Sampling Systems
Container, conical, with screw cap, Corning® Gosselin™
Supplier: Corning
Filter plates, 96-well, AcroPrep™
Supplier: Cytiva (Formerly Pall Lab)
Transfer pipettes
Supplier: RATIOLAB
[EN]LID FOR ICE BUCKET 2.5 / 4.5 L-GRN 1 * 1 KS
Supplier: VWR Collection
[EN]VIAL 167ML ACTIVE 900 M-3003-220 1 * 1.800 KS
Supplier: Capitol Europe