Masterflex® Fitting, PVDF, Tee, Hose Barb Unions, 1/16" ID; 10/PK
Katalogové číslo MFLX40615-54
Dodavatel: Avantor Fluid Handling
O tomto produktu
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Barbed tee fittings are designed for quick and easy assembly. Combine these simple, economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Velikost ozubu/vnit. Ø hadičky (port 1):¹⁄₁₆"
- Velikost ozubu/vnit. Ø hadičky (port 2):¹⁄₁₆"
- Typ připojení:Barbed Union
- Flow pattern:T
- Popis:Fitting, PVDF, Tee, Hose Barb Unions, ¹⁄₁₆" ID; 10/PK
- Environmentally Preferable:
- Sterilizace:Autoclave or ethylene oxide (ETO)
- Bal.:10
- Barb size/tubing int.Ø (port 3):¹⁄₁₆"
- Fitting material:PVDF
Frequently Bought Together