[EN]FILTER DISCS2.1MM ID0.2M QUICK CHANG 1 * 5 KS
Katalogové číslo HEWL5067-1610
Dodavatel: Agilent
Frequently Bought Together
Aceton ≥99.5%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
VWR®, Crimp Neck Vials ND11
Dodavatel: VWR Collection
VWR®, Short Thread Vials ND9
Dodavatel: VWR Collection
Aceton ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Dodavatel: VWR Chemicals
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
VWR®, Nitrile Gloves
Dodavatel: VWR Collection
VWR®, Ubrousky čisticí
Dodavatel: VWR Collection
Acetonitril ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
Hydrogenuhličitan amonný 99% for analysis
Dodavatel: Thermo Fisher Scientific
1290 valve head capillaries
Dodavatel: Agilent