Masterflex® I/P® Easy-Load® Replacement Rotor and Bearing Assembly for 77602-02 Pump Head
Catalog # MFLX77602-28
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® I/P® Easy-Load® Pump Head with these replacement parts.
- Suitable for use with I/P® Easy-Load® pump heads
Replacement parts to keep your pump head operating even longer.
- Pump series:I/P
- Popis:I/P® Easy-Load® Replacement Rotor and Bearing Assembly for 77602-02 Pump Head
- Environmentally Preferable:
- Záruka:3 Months
- Bal.:1
- Housing material:PPS
- Rotor material:Coled-Rolled Steel
Frequently Bought Together