Masterflex® Fitting, Natural Polypropylene, Tee, Hose Barb to Thread Adapter, 3/4" ID x 3/4" ID x 3/4" NPT(M); 5/PK
Katalogové číslo MFLX40610-87
Dodavatel: Avantor Fluid Handling
CE certificate
O tomto produktu
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
- Velikost ozubu/vnit. Ø hadičky (port 2):¾"
- Typ připojení:Thread to Barb
- Flow pattern:T
- Popis:Fitting, Natural Polypropylene, Tee, Hose Barb to Thread Adapter, ¾" ID × ¾" ID × ¾" NPT(M); 5/PK
- Sekvence:13
- Environmentally Preferable:
- Model:40610-87
- Orifice dimensions:0,5625"
- Sterilizace:Ethylene oxide (ETO) or gamma
- Záruka:Out of box
- Bal.:5
- Fitting material:Polypropylene
Frequently Bought Together