Masterflex® Fitting, 316 Stainless Steel, Tee, Hosebarb to Thread, 1/4" ID x 1/4" ID x 3/8" NPT(M)
Katalogové číslo MFLX31209-29
Dodavatel: Avantor Fluid Handling
O tomto produktu
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/4" ID x 1/4" ID x 3/8" NPT(M)
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
- Velikost ozubu/vnit. Ø hadičky (port 2):1/4"
- Typ připojení:Thread to Barb
- Flow pattern:T
- Popis:Fitting, 316 Stainless Steel, Tee, Hosebarb to Thread, 1/4" ID x 1/4" ID x 3/8" NPT(M)
- Sekvence:12
- Environmentally Preferable:
- Model:31209-29
- Orifice dimensions:0,1875"
- Záruka:1 year
- Bal.:1
- Barb size/tubing int.Ø (port 3):1/4"
- Fitting material:Stainless Steel 316
Frequently Bought Together