Masterflex® Fitting, 303 Stainless Steel, Tee, Hosebarb Reducer, 3/8" ID x 3/8" ID x 1/8" ID
Catalog # MFLX31208-47
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/8" ID x 3/8" ID x 1/8" ID
Barbed fittings are designed for quick and easy assembly without tools. Combine these simple and economical fittings with our flexible tubing for use in your low-pressure applications (generally less than 30 psi).
- Velikost ozubu/vnit. Ø hadičky (port 1):3/8"
- Velikost ozubu/vnit. Ø hadičky (port 2):3/8"
- Typ připojení:Barbed Reducer
- Flow pattern:T
- Popis:Fitting, 303 Stainless Steel, Tee, Hosebarb Reducer, 3/8" ID x 3/8" ID x 1/8" ID
- Sekvence:16
- Environmentally Preferable:
- Model:31208-47
- Orifice dimensions:0,09375"
- Sterilizace:Autoclave, ethylene oxide (ETO), or gamma
- Záruka:1 year
- Bal.:1
- Barb size/tubing int.Ø (port 3):1/8"
- Fitting material:Stainless Steel 316
Frequently Bought Together