Masterflex® Fittings, HDPE, Elbow, Hose Barb Union, 3/16" ID; 100/PK
Catalog # MFLX30626-72
Supplier: Avantor Fluid Handling
CE certificate
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/16" ID
Create 90° bends in flexible tubing—without the kinks!
- Velikost ozubu/vnit. Ø hadičky (port 1):3/16"
- Velikost ozubu/vnit. Ø hadičky (port 2):3/16"
- Typ připojení:Barbed Union
- Flow pattern:Elbow
- Popis:Fittings, HDPE, Elbow, Hose Barb Union, 3/16" ID; 100/PK
- Sekvence:08
- Environmentally Preferable:
- Model:30626-72
- Orifice dimensions:0,09375"
- Sterilizace:Ethylene oxide (ETO) only
- Bal.:100
- Fitting material:HDPE
Frequently Bought Together