Masterflex® Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 5/32" ID x 1/4" NPT(M); 100/PK
Catalog # MFLX30625-40
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 5/32" ID x 1/4" NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Velikost ozubu/vnit. Ø hadičky (port 2):5/32"
- Typ připojení:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):1/4"
- Popis:Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 5/32" ID x 1/4" NPT(M); 100/PK
- Sekvence:37
- Environmentally Preferable:
- Model:30625-40
- No. of ports:2
- Sterilizace:Ethylene oxide (ETO) only
- Bal.:100
- Fitting material:Nylon
Frequently Bought Together