Masterflex® Fitting, PVDF, Straight, Hose Barb Reducer, 1/2" ID x 3/8" ID; 10/PK
Katalogové číslo MFLX30616-24
Dodavatel: Avantor Fluid Handling
CE certificate
O tomto produktu
No tools needed for assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/2" ID x 3/8" ID
Hose barb fittings make tubing connections quick and easy, requiring no tools for installation. Select the fitting based on the Masterflex tubing size. Fittings are available in a variety of materials to suit different applications and budgets.
- Velikost ozubu/vnit. Ø hadičky (port 1):0,5"
- Velikost ozubu/vnit. Ø hadičky (port 2):0,375"
- Typ připojení:Barb
- Flow pattern:Straight
- Popis:Kynar REDUCER,I/P 82/88 & BT 88,10/P
- Sekvence:22
- Environmentally Preferable:
- Model:30616-24
- Sterilizace:Autoclave or ethylene oxide (ETO)
- Bal.:10
- Fitting material:PVDF
Frequently Bought Together