Smart Gasket Sanitary Sampling Gasket, Silicone, 4 Port, 2" Tri-Clamp
Catalog # MFLX30545-16
Supplier: Avantor Fluid Handling
Specifications
See all product configurations- Catalog No:
- MFLX30545-16
- MFLX30544-06
- MFLX30543-14
- MFLX30544-05
- MFLX30543-08
- MFLX30543-13
- MFLX30543-05
- MFLX30543-16
- MFLX30544-00
- MFLX30544-04
- MFLX30543-18
- MFLX30542-10
- MFLX30542-09
- MFLX30542-18
- MFLX30542-17
- MFLX30542-00
- MFLX30543-00
- MFLX30543-04
- MFLX30542-02
- MFLX30542-16
- MFLX30542-01
- MFLX30544-14
- MFLX30543-12
- MFLX30542-04
- MFLX30542-05
- MFLX30542-08
- MFLX30542-06
- MFLX30542-14
- MFLX30542-12
- MFLX30542-13
- MFLX30545-13
- MFLX30545-12
- MFLX30544-12
- MFLX30544-16
- MFLX30545-14
- Typ připojení:
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- No. of ports:
- 4
- 3
- 2
- 3
- 2
- 2
- 2
- 2
- 3
- 3
- 2
- 1
- 1
- 1
- 1
- 1
- 2
- 2
- 1
- 1
- 1
- 3
- 2
- 1
- 1
- 1
- 1
- 1
- 1
- 1
- 4
- 4
- 3
- 3
- 4
- Záruka:
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- 1 year
- Currently Viewing
Specifications
About this item
Smart Gasket lets you take samples or readings from a sanitary line.
- Measure temperature, take a sample, or dose chemicals into your process line
- Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
- Sanitary clamp size (Port 1):2"
- Typ připojení:Sanitary
- Popis:2" Tri-Clamp, 4 Port, Silicone, Each
- Sekvence:15
- Environmentally Preferable:
- Bal.:1
- Fitting material:Platinum-cured silicone
- Sanitary gasket size:2"
- No. of ports:4
- Záruka:1 year
Frequently Bought Together