Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
19, 3/4" Tri-Clamp, 3 Port, Viton, Each, Viton, 3/4", 3/4"
Katalogové číslo MFLX30544-06
Dodavatel: Avantor Fluid Handling
Masterflex® Smart Gasket® Sanitary Sampling Gaskets, Avantor®
Katalogové číslo MFLX30544-06
Dodavatel: Avantor Fluid Handling
Přejít na:
Product Details & Documents
Smart Gasket lets you take samples or readings from a sanitary line.
- Measure temperature, take a sample, or dose chemicals into your process line
- Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
Documents
Certifikáty
Hledat podle čísla šaržeSpecifikace
- Sanitary clamp size (Port 1):3/4"
- Popis:3/4" Tri-Clamp, 3 Port, Viton, Each
- Sekvence:19
- Fitting material:Viton
- Sanitary gasket size:3/4"
- Cat. No.:MFLX30544-06
- Typ připojení:Sanitary
- Záruka:1 year
- No. of ports:3