Smart Gasket 1" Tri-Clamp Gasket, 1 Port, EPDM; 1/Each
MFLX30542-09
Smart Gasket lets you take samples or readings from a sanitary line.
- Measure temperature, take a sample, or dose chemicals into your process line
- Install in between two sanitary fittings
The Smart Gasket is used as a standard sanitary gaskets with standard Tri-clamp® fittings. The gaskets have molded ports that allow either a thermocouple or sampling tubes to be inserted into the line. Order the appropriate Smart Gasket clamp separately.
- Sanitary clamp size (Port 1):1"
- Typ připojení:Sanitary
- Popis:1" Tri-Clamp, 1 Port, EPDM, Each
- Sekvence:05
- Environmentally Preferable:
- Bal.:1
- Fitting material:EPDM
- Sanitary gasket size:1"
- No. of ports:1
- Záruka:1 year
Frequently Bought Together