[EN]SKILLPAK DEAE-650C 5 ML COL. 8.0X100 1 * 1 KS
Katalogové číslo TOYO0045287
Dodavatel: TOSOH Bioscience
[EN]SKILLPAK DEAE-650C 5 ML COL. 8.0X100 1 * 1 KS
Katalogové číslo TOYO0045287
Dodavatel: TOSOH Bioscience
Frequently Bought Together
Avantor® ACE® AQ, HPLC Columns, 5 µm
Dodavatel: Avantor
VWR®, Screw Neck Vials ND24
Dodavatel: VWR Collection
Avantor® ACE® C8, HPLC Columns, 5 µm
Dodavatel: Avantor
Chromatogram immersion device for TLC plates
Dodavatel: CAMAG
VWR® Hichrom Precolumn Filters
Dodavatel: Avantor
Screw caps and septa, 9 mm
Dodavatel: THERMO NATIONAL SCIENTIFIC
[EN]2ML LA VIAL, CLEAR, PTFE/SILICONE 1 * 100 KS
Dodavatel: Merck
Tubing
Dodavatel: S.C.A.T.
Consumables for Agilent Technologies autosamplers
Dodavatel: Trajan Scientific and Medical
Tubing, PTFE
Dodavatel: HAMILTON BONADUZ
CRS ONE O-Rings for Inlet Liner Seals
Dodavatel: Trajan Scientific and Medical
Micro extraction by packed sorbent (MEPS™)
Dodavatel: Trajan Scientific and Medical