Suction Line Strainer, PVC, 1/2" NPT(F), 20 Mesh (stainless steel)
Katalogové číslo MFLX74604-20
Dodavatel: Avantor Fluid Handling
O tomto produktu
Protect pump from sediments filtering into suction line.
- PVC (polyvinyl chloride) or Nylon base material of construction
- 20, 40, 80 mesh sizes available
- Easily removable for cleaning or replacing
These suction line pump filter strainers are ideal for keeping sediment and debris out of your pump for optimal performance. Choose from PVC (polyvinyl chloride) or nylon base material in a wide range of mesh sizes for any application.
- Popis:Suction Line Strainer, PVC, 1/2" NPT(F), 20 Mesh (stainless steel)
- Environmentally Preferable:
- Máčené materiály:PVC and Stainless Steel
- Outlet connection:0,5 NPT(F)
- Záruka:1 year
- Bal.:1
- Inlet connection:0,5 NPT(F)
Frequently Bought Together