GF Signet 3-2552-21-A-12 Magmeter Flow Sensor, 9.3" Depth, 25' Cable, 4-20 mA
Katalogové číslo MFLX32486-45
Dodavatel: Avantor Fluid Handling
Specifikace
Zobrazit všechny konfigurace produktu- Typ nastavení:
- On/Off
- Záruka:
- 2 years
- See message
- 1 year part and 2 years labor
- 1 year part and 2 years labor
- Právě zobrazeno
Specifikace
O tomto produktu
Optimize performance of your GF+ Signet Transmitter/Controller.
- Choose from a wide range of replacement parts and accessories
Use modules, sensors, and RC filters to expand the controller's capabilities. Adapt your controller to your existing system with a variety of mounting options.
- Popis:Magmeter Flow Sensor, 9,3" Depth, 25' Cable, 4-20 mA
- Environmentally Preferable:
- Záruka:2 years
- Bal.:1
Frequently Bought Together