Compression Fitting, Male Pipe Adapter (Straight), 316 SS, 1/4" OD x 1/2" NPT
Katalogové číslo MFLX31406-37
Dodavatel: Avantor Fluid Handling
CE certificate
O tomto produktu
Excellent choice to connect rigid tubing.
- Great chemical resistance
- Easy to assemble the fitting to the tubing
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through the included sealed ferrule and using the nut to compress down to form a secure seal.
Very good chemical resistance. Pressure limitation determined by the tubing, not the fitting. Usable from -40 to 200ºF (-40 to 93ºC). Sterilize by either ethylene oxide, autoclave, or gamma irradiation.
- Typ připojení:Compression to Thread
- Flow pattern:Straight
- Thread size NPT(M) (port 2):1/2"
- Popis:Compression Fitting, Male Pipe Adapter (Straight), 316 SS, 1/4" OD×1/2" NPT
- Environmentally Preferable:
- Sterilizace:Autoclave, ethylene oxide (ETO), or gamma
- Záruka:LTD life time
- Bal.:1
- Fitting material:Stainless Steel 316
Frequently Bought Together