Masterflex® I/P® Easy-Load® Pump Head, Replacement Rotor and Bearing Assembly, PPS Housing, CRS Rotor
Katalogové číslo MFLX77602-08
Dodavatel: Avantor Fluid Handling
O tomto produktu
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for Easy-Load® Pump Head 77602-00
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:I/P
- Popis:Easy-Load® Pump Head, Replacement Rotor and Bearing Assembly, PPS Housing, CRS Rotor; 1/Ea
- Sekvence:43
- Environmentally Preferable:
- Model:77602-08
- Open-head sensor:No
- Záruka:3 Months
- Bal.:1
- Housing material:PPS
- Rotor material:Cold-rolled steel (CRS)
Frequently Bought Together