Masterflex® I/P® Replacement Main Body, PPS Housing, CRS Rotor
Catalog # MFLX77602-07
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® I/P® Easy-Load® Pump Head with these replacement parts.
- Suitable for use with pump heads 77601-60 and 77601-62
Keep your pump head operating at its best with these replacement main body and occusion bed assemblies.
- Pump series:I/P
- Popis:Replacement Main Body, PPS Housing, CRS Rotor
- Environmentally Preferable:
- Záruka:3 Months
- Bal.:1
- Housing material:PPS
- Rotor material:Cold-Rolled Steel
Frequently Bought Together