Masterflex® Fitting, Polypropylene, Straight, Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 1/8"; 25/PK
Katalogové číslo MFLX45508-34
Dodavatel: Avantor Fluid Handling
CE certificate
O tomto produktu
Ensure your process integrity with full material traceability.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: Female Luer Bulkhead to Hose Barb, 1/4-28 UNF x 1/8"
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
- Velikost ozubu/vnit. Ø hadičky (port 1):1/8"
- Typ připojení:Female Luer to Barb
- Flow pattern:Straight
- Popis:Fitting, Polypropylene, Straight, Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 1/8"; 25/PK
- Sekvence:03
- Environmentally Preferable:
- Connection type (port 1):Female luer
- Connection type (port 2):Hose barb
- No. of ports:2
- Bal.:25
- Fitting material:Polypropylene
Frequently Bought Together