O tomto produktu
Ensure your process integrity with full material traceability.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Designed for quick and easy assembly with flexible tubing
- Connection Type: Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 1/16" ID
These luer fittings are ideal for a variety of applications, including syringes or small tubing. Barbed adapters feature three styles of barbs: standard, wide-bore, and low-profile. Wide-bore barbs offer enhanced fluid flow through the fitting and are usable with any flexible tubing. Low-profile barbs are ideal for use with semi-rigid tubing, such as nylon, polyethylene, or polypropylene. Manufactured in compliance with ISO 594-1 standards, these fittings are cleaned, inspected and fully traceable. All parts meet tight dimensional tolerances for increased reliability and process purity.
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
- Velikost ozubu/vnit. Ø hadičky (port 1):1/16"
- Typ připojení:Female Luer to Barb
- Flow pattern:Straight
- Popis:Fitting, Polycarbonate, Straight, Female Luer Bulkhead to Hose Barb Adapter, 1/4-28 UNF x 1/16" ID; 25/PK
- Sekvence:12
- Environmentally Preferable:
- Model:45501-30
- Connection type (port 1):Female luer
- No. of ports:3
- Záruka:3 Months
- Bal.:25
- Fitting material:Polycarbonate