Masterflex® Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 1/2" ID x 1/2" NPT(M)
Katalogové číslo MFLX34030-83
Dodavatel: Avantor Fluid Handling
CE certificate
O tomto produktu
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1/2" ID x 1/2" NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Velikost ozubu/vnit. Ø hadičky (port 2):1/2"
- Typ připojení:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):1/2"
- Popis:Fitting, PTFE, Straight, Hose Barb to Threaded Adapter, 1/2" ID x 1/2" NPT(M)
- Sekvence:11
- Environmentally Preferable:
- No. of ports:2
- Sterilizace:Autoclave or ethylene oxide (ETO)
- Záruka:2 years
- Bal.:1
- Fitting material:PTFE
Frequently Bought Together