Masterflex® Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1 1/4" ID x 1 1/2" NPT(M); 5/PK
Katalogové číslo MFLX31408-17
Dodavatel: Avantor Fluid Handling
O tomto produktu
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 1 1/4" ID x 1 1/2" NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Velikost ozubu/vnit. Ø hadičky (port 2):1-1/4"
- Typ připojení:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):1-1/2"
- Popis:Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1 1/4" ID x 1-1/2" NPT(M); 5/PK
- Sekvence:52
- Environmentally Preferable:
- Model:31408-17
- No. of ports:2
- Orifice dimensions:15,08 mm (0,59375")
- Sterilizace:Ethylene oxide (ETO) only
- Záruka:1 year
- Bal.:5
- Fitting material:Nylon
Frequently Bought Together