Masterflex® Fitting, 316 Stainless Steel, Straight, Hosebarb to Thread, 3/4" ID x 1/2" NPT(M)
Catalog # MFLX31210-34
Supplier: Avantor Fluid Handling
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: 3/4" ID x 1/2" NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Velikost ozubu/vnit. Ø hadičky (port 2):3/4"
- Typ připojení:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):1/2"
- Popis:Fitting, 316 Stainless Steel, Straight, Hosebarb to Thread, 3/4" ID x 1/2" NPT(M)
- Sekvence:25
- Environmentally Preferable:
- Model:31210-34
- No. of ports:2
- Orifice dimensions:13,49 mm (0,53125")
- Záruka:1 year
- Bal.:1
- Fitting material:Stainless Steel 316
Frequently Bought Together