Masterflex® Fitting, Polypropylene, Straight, Female Luer to Hosebarb Adapters, 1/8" ID; 25/PK
Catalog # MFLX30800-08
Supplier: Avantor Fluid Handling
CE certificate
About this item
Easily transition to a luer fitting.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: Female luer to 1/8" hose barb with 0.44" diameter orifice
Manufactured of animal-free natural polypropylene, these luers meet Class VI specifications. Luers offer superior chemical resistance and are suited for ETO and gamma sterilization.
- Velikost ozubu/vnit. Ø hadičky (port 1):⅛"
- Typ připojení:Female Luer to Barb
- Flow pattern:Straight
- Popis:Fitting, Polypropylene, Straight, Female Luer to Hosebarb Adapters, ⅛" ID; 25/PK
- Sekvence:04
- Environmentally Preferable:
- Model:30800-08
- Connection type (port 1):Female luer
- Connection type (port 2):Hose barb
- No. of ports:2
- Záruka:3 Months
- Bal.:25
- Fitting material:Polypropylene
Frequently Bought Together