Masterflex® Fittings, HDPE, Elbow, Hose Barb Union, 3/8" ID; 10/PK
Catalog # MFLX30622-78
Supplier: Avantor Fluid Handling
CE certificate
About this item
Quick and easy assembly without tools.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Create 90° bends in flexible tubing—without the kinks!
- Velikost ozubu/vnit. Ø hadičky (port 1):3/8"
- Velikost ozubu/vnit. Ø hadičky (port 2):3/8"
- Typ připojení:Barbed Union
- Flow pattern:Elbow
- Popis:Fittings, HDPE, Elbow, Hose Barb Union, 3/8" ID; 10/PK
- Sekvence:06
- Environmentally Preferable:
- Model:30622-78
- Orifice dimensions:0,21875"
- Sterilizace:Ethylene oxide (ETO) only
- Bal.:10
- Fitting material:HDPE
Frequently Bought Together