Masterflex® Smart Gasket® Hole Plug for System, Stainless Steel; 1/EA
Katalogové číslo MFLX30545-98
Dodavatel: Avantor Fluid Handling
O tomto produktu
Optimize performance of your Masterflex® Smart Gasket® Sanitary Gasket system.
- Choose from a wide range of replacement parts and accessories
Smart Gaskets allow you to measure temperature, take samples, or dose chemicals in your process line. Selected accessories include thermocouple probes and wire, port plugs for the Smart Gasket, and a sampling tube.
- Typ připojení:Sanitary
- Popis:Hole Plug for System, Stainless Steel; 1/EA
- Sekvence:01
- Environmentally Preferable:
- Bal.:1
- Fitting material:Stainless Steel
- Záruka:1 year
Frequently Bought Together