Masterflex® Footswitch, On/Off Operation, Wash-Down; 18-Pin Round, 6-ft Cable
Catalog # MFLX07595-43
Supplier: Avantor Fluid Handling
About this item
Easily control your Masterflex® drive with compatible footswitch.
- Choose footswitch with the correct connection for your Masterflex® pump
Add convenience to repetitive dispensing with a foot-operated on/off switch. Keep hands free to efficiently move containers during routine fill applications or to keep containers stable during the filling process.
- Popis:Wash-Down On/Off Footswitch; 18-Pin Round, 6-ft Cable
- Environmentally Preferable:
- Záruka:2 years
- Bal.:6 Feet
Frequently Bought Together