Masterflex® L/S® Easy-Load® Pump Head, Replacement Rotor and Bearing Assembly, White PPS Housing, CRS Rotor, for 07516-00
Katalogové číslo MFLX07516-08
Dodavatel: Avantor Fluid Handling
O tomto produktu
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
- Suitable for Easy-Load® Pump Heads 07516-00 and 07518-00
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- Popis:Easy-Load® Pump Head, Replacement Rotor and Bearing Assembly, White PPS Housing, CRS Rotor; 1/Ea
- Sekvence:38
- Environmentally Preferable:
- Model:07516-08
- Open-head sensor:No
- Záruka:2 years
- Bal.:1
- Housing material:PPS
- Rotor material:Cold-rolled steel (CRS)
Frequently Bought Together