Masterflex® L/S® Easy-Load® Pump Head Replacement Rotor/Bearing Assembly for 07514-10 and -30
Katalogové číslo MFLX07514-18
Dodavatel: Avantor Fluid Handling
1 year warranty
O tomto produktu
Get the most our of your Masterflex® Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® and I/P® pump head
Replacement parts to keep your pump head operating even longer. Two end bell assemblies are required for a complete head assembly.
- Pump series:L/S
- Popis:L/S® Easy-Load® Pump Head Replacement Rotor/Bearing Assembly for 07514-10 and -30
- Sekvence:36
- Environmentally Preferable:
- Model:07514-18
- Open-head sensor:No
- Tubing sizes accepted:L/S® 13, L/S 14, L/S 16, L/S 25, L/S 17, L/S 18
- Záruka:1 year
- Bal.:1
- Válečky:4
- Rotor material:Stainless Steel
Frequently Bought Together