Masterflex® Sanitary Clamp with White Knob, Nylon, 1/2" to 3/4"; 1/EA
Katalogové číslo MFLX30508-21
Dodavatel: Avantor Fluid Handling
CE certificate
O tomto produktu
Compatible with metal and plastic Tri-Clamp® fittings.
- Excellent chemical resistance; low leachable characteristics
- Autoclavable
- Accepts 1/2" to 3/4" tubing
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.
- Typ připojení:Sanitary Clamp
- Flow pattern:Straight
- Sanitary clamp size (Port 1):3/4"
- Popis:Sanitary Clamp with White Knob, Nylon, 1/2" to 3/4"; 1/EA
- Sekvence:01
- Environmentally Preferable:
- Model:30508-21
- Sterilizace:Autoclave, Gamma
- Záruka:Manufacturer defect
- Bal.:1
- Fitting material:Nylon
- Sanitary gasket size:3/4"
Frequently Bought Together